Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab5a Antibody (3A4) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4998232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104340SS 0.02 mL
NBP104340 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP104340SS Supplier Novus Biologicals Supplier No. NBP104340SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab5a Monoclonal specifically detects Rab5a in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rab5a
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3A4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Gene Accession No. NP_004153
Gene Alias RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A
Gene Symbols RAB5A
Host Species Mouse
Immunogen Recombinant human Rab5a (1-215aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Endosome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5868
Test Specificity Anti-human Rab5a mAb, clone 3A4, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Rab5a protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.