Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab5b Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299625JF549
Description
Rab5b Polyclonal antibody specifically detects Rab5b in Human samples. It is validated for Western BlotSpecifications
Rab5b | |
Polyclonal | |
Western Blot | |
RAB5B, member RAS oncogene family, ras-related protein Rab-5B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Rab5b. | |
0.1 mL | |
Endosome Markers, Signal Transduction | |
5869 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction