Learn More
Invitrogen™ RAB5B Monoclonal Antibody (1615CT668.28.45)

Description
RAB5B Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RAB5B |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1615CT668.28.45 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RAB5B |
| Gene Accession No. | P61020, P61021 |
| Gene Alias | C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.