Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578144 Supplier Invitrogen™ Supplier No. PA578144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HeLa, HeLa membrane extract Predicted Reactivity: Mouse (100%), Rat (98%), Chicken (96%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RAB5B
Gene Accession No. P61020
Gene Alias C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978
Gene Symbols RAB5B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Rab5b. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5869
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.