Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110261
Description
Immunogen sequence: YDITNQETFA RAKTWVKELQ RQASPSIVIA LAGNKADLAN KRMVEYEEAQ AYADDNSLLF METSAKTAMN VNDLFLAIAK KLPKSEPQNL GGAAGRSRGV D.
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.
Specifications
| RAB5B | |
| Polyclonal | |
| Unconjugated | |
| RAB5B | |
| C030027M18Rik; RAB5A member RAS oncogene family; RAB5B; RAB5B, member RAS oncogene family; ras-related protein Rab-5B; zgc:76978 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19344, 288779, 5869 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.34 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P61020, P61021 | |
| RAB5B | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human RAB5B (NP_0028591). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction