Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB6A Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP325946FR
Description
RAB6A Polyclonal antibody specifically detects RAB6A in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
RAB6A | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
GTP-binding protein RAB6A', GTP-binding protein RAB6B, Oncogene RAB6, Rab GTPase, Rab-6, RAB6, member RAS oncogene family, RAB6A', Rab6A variant 3, RAB6A, member RAS oncogene family, RAB6B, RAB6rab-6, ras-related protein Rab-6A | |
Rabbits immunized with purified, recombinant Human RAB6A (Accession#: P20340-2, Met1-Cys208) | |
0.1 mL | |
Signal Transduction | |
5870 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction