Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588341 Supplier Invitrogen™ Supplier No. PA588341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSTGGDFGNP LRKFKLVFLG EQSVGKTSLI TRFMYDSFDN TYQATIGIDF LSKTMYLEDR TIRLQLWDTA GQERFRSLIP SYIRDSAAAV VVYDITNVNS FQQTTKWIDD VRTERGSDVI IMLVGNKTDL ADKRQVSIEE GERKAKELNV MFIETSAKAG YNVKQLFRRV AAALPGMEST QDRSREDMID IKLEKPQEQP; Positive Samples: Mouse brain; Cellular Location: Golgi apparatus, Golgi apparatus membrane, Lipid-anchor.

Rab proteins are also an integral part of endocytic pathways. Rab 6A is a ubiquitously expressed member of the Rab family of proteins and localizes to the Golgi membrane where it regulates retrograde transport from the late endosomes via the Golgi to endoplasmic reticulum (ER) pathway. Three isoforms exist due to alternative splicing events, namely the ubiquitously expressed isoforms Rab 6A' and Rab 6A, and the brain-specific isoform Rab 6B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB6A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAB6A
Gene Accession No. P35279
Gene Alias 2610028L11Rik; AA419671; MNCb-1660; Rab GTPase; Rab6; rab-6; RAB6, member RAS oncogene family; RAB6A; RAB6A, member RAS oncogene family; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6A; RCO4-3
Gene Symbols RAB6A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAB6A (NP_0028602).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19346
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.