Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB6B Monoclonal Antibody (6D12E4)
GREENER_CHOICE

Catalog No. PIMA531882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531882 Supplier Invitrogen™ Supplier No. MA531882
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31882 has been tested in indirect ELISA.

The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB6B
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6D12E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Rab6b
Gene Accession No. Q9NRW1
Gene Alias 9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6B; small GTPase RAB6B; small GTP-binding protein
Gene Symbols Rab6b
Host Species Mouse
Immunogen Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363123, 51560
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.