Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB6B Monoclonal Antibody (6D12G9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531881
Description
MA5-31881 has been tested in indirect ELISA.
The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
Specifications
RAB6B | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9NRW1 | |
Rab6b | |
Purified recombinant fragment of human Rab6b (AA: 95-208) expressed in E. coli. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
6D12G9 | |
Unconjugated | |
Rab6b | |
9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6B; small GTPase RAB6B; small GTP-binding protein | |
Mouse | |
Protein G | |
RUO | |
363123, 51560 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction