Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571330
Description
RAB6B Polyclonal Antibody for Western Blot
The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
Specifications
RAB6B | |
Polyclonal | |
Unconjugated | |
Rab6b | |
9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6B; small GTPase RAB6B; small GTP-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
51560 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NRW1 | |
Rab6b | |
synthetic peptide directed towards the C-terminal region of human RAB6B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction