Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598909
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598909 Supplier Invitrogen™ Supplier No. PA598909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB6B Polyclonal Antibody for Western Blot, IHC (P), ELISA

The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB6B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Rab6b
Gene Accession No. P61294, Q9NRW1
Gene Alias 9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6B; small GTPase RAB6B; small GTP-binding protein
Gene Symbols Rab6b
Host Species Rabbit
Immunogen Recombinant Human Ras-related protein Rab-6B protein (2-208AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270192, 51560
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.