Learn More
Invitrogen™ RAB6C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114158
Description
Immunogen sequence: DVIITLVGNR TDLADKRQVS VEEGERKAKG LNVTFIETRA KAGYNVKQLF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 92%; Goat: 85%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Yeast: 92%; Zebrafish: 92%.
May be involved in the regulation of centrosome duplication and cell cycle progression.
Specifications
RAB6C | |
Polyclonal | |
Unconjugated | |
RAB6C | |
RAB6C; RAB6C, member RAS oncogene family; rab6-like protein WTH3; ras-related protein Rab-6C; WTH3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84084 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0N0 | |
RAB6C | |
Synthetic peptide directed towards the middle region of human RAB6C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.