Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB6C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101829
Description
Antibody detects endogenous levels of total RAB6C.
May be involved in the regulation of centrosome duplication and cell cycle progression.
Specifications
RAB6C | |
Polyclonal | |
Unconjugated | |
RAB6C | |
RAB6C; RAB6C, member RAS oncogene family; rab6-like protein WTH3; ras-related protein Rab-6C; WTH3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84084 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H0N0 | |
RAB6C | |
A synthesized peptide derived from human RAB6C(Accession Q9H0N0), corresponding to amino acid residues K158-C208. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction