Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB6IP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RAB6IP1 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RAB6IP1 Polyclonal specifically detects RAB6IP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RAB6IP1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DENN domain-containing protein 5A, DENN/MADD domain containing 5A, FLJ22354, FLJ33829, FLJ43455, KIAA1091Rab6-interacting protein 1, RAB6 interacting protein 1, RAB6IP1 | |
DENND5A | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Rabbit | |
Human | |
Q6IQ26 | |
23258 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MHNAEYDVLHAPPADDRDQSSMEDGEDTPVTKLQR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title