Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552369
Description
Immunogen sequence: CVLVFDVTAP NTFKTLDSWR DEFLIQASPR DPENFPFVVL GNKIDLENRQ VATKRAQAWC YSKNNIPYFE TSAKEAINVE QAFQTIARNA LKQETEVELY NEFPEPIKLD KNDRAK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
RAB7 | |
Polyclonal | |
Unconjugated | |
RAB7A | |
GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110315128, 19349, 29448, 7879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09527, P51149, P51150 | |
Rab7, RAB7A | |
Recombinant protein corresponding to Human RAB7. Recombinant protein control fragment (Product #RP-89503). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction