Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB7A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109239
Description
Positive Control: MCF-7, A431, SiHa, rat brain tissue, human tonsil tissue, human liver tissue, human colon cancer tissue, mouse colon tissue.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
Specifications
RAB7A | |
Polyclonal | |
Unconjugated | |
RAB7A | |
GTP-binding protein (rab7); hypothetical protein LOC509970; PRO2706; RAB7; rab-7; RAB7, member RAS oncogene family; Rab7a; RAB7A, member RAS oncogene family; Ras-associated protein RAB7; ras-related protein BRL-RAS; Ras-related protein p23; ras-related protein Rab-7a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7879 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P51149 | |
RAB7A | |
Recombinant protein within Human RAB7 aa 43-200. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction