Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549264
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549264 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549264 Supplier Thermo Scientific Supplier No. PA549264
Add to Cart
Add to Cart

RAB7B Polyclonal Antibody

RAB7B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB7B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB7B
Gene Accession No. Q96AH8
Gene Alias 5430435G22Rik; RAB7; Rab7b; RAB7B, member RAS oncogene family; Ras-related protein Rab-7; ras-related protein Rab-7b; RGD1562617; RIKEN cDNA 5430435G22 gene
Gene Symbols RAB7B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 168-197 amino acids from the C-terminal region of human RAB7B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338382
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.