Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110262 Supplier Invitrogen™ Supplier No. PA5110262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence of this protein is as follows: MNPRKKVDLK LIIVGAIGVG KTSLLHQYVH KTFYEEYQTT LGASILSKII ILGDTTLKLQ IWDTGGQERF RSMVSTFYKG SDGCILAFDV TDLESFEALD IWRGDVLAKI VPMEQSYPMV LLGNKIDLAD RKVPQEVAQG WCREKDIPYF EVSAKNDINV VQAFEMLASR ALSRYQSILE NHLTESIKLS PDQSRSRCC.

Controls vesicular trafficking from endosomes to the trans-Golgi network (TGN). Acts as a negative regulator of TLR9 signaling and can suppress TLR9-triggered TNFA, IL6, and IFNB production in macrophages by promoting TLR9 lysosomal degradation. Also negatively regulates TLR4 signaling in macrophages by promoting lysosomal degradation of TLR4. Promotes megakaryocytic differentiation by increasing NF-kappa-B-dependent IL6 production and subsequently enhancing the association of STAT3 with GATA1. Not involved in the regulation of the EGF- and EGFR degradation pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB7B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RAB7B
Gene Accession No. Q8VEA8, Q96AH8
Gene Alias 5430435G22Rik; RAB7; Rab7b; RAB7B, member RAS oncogene family; Ras-related protein Rab-7; ras-related protein Rab-7b; RGD1562617; RIKEN cDNA 5430435G22 gene
Gene Symbols RAB7B
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226421, 338382
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.