Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB8A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179960
Description
RAB8A Polyclonal antibody specifically detects RAB8A in Human samples. It is validated for Western Blot.Specifications
RAB8A | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
mel transforming oncogene (derived from cell line NK14), mel transforming oncogene (derived from cell line NK14)- RAB8 homolog, MELras-associated protein RAB8, Oncogene c-mel, RAB8A, member RAS oncogene family, RAB8mel transforming oncogene (RAB8 homolog), ras-related protein Rab-8A | |
Rabbit | |
24 kDa | |
100 μL | |
Primary | |
Mouse: 85%; Rat: 85%; Zebrafish: 85%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Sheep, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_005361 | |
RAB8A | |
Synthetic peptide directed towards the middle region of human RAB8AThe immunogen for this antibody is RAB8A. Peptide sequence GIKFMETSAKANINVENAFFTLARDIKAKMDKKLEGNSPQGSNQGVKITP. | |
Affinity purified | |
RUO | |
4218 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction