Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB8A Monoclonal Antibody (261CT1.3.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524717
Description
RAB8A Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the RAS superfamily which are small GTP/GDP-binding proteins with an average size of 200 amino acids. The RAS-related proteins of the RAB/YPT family may play a role in the transport of proteins from the endoplasmic reticulum to the Golgi and the plasma membrane. This protein shares 97%, 96%, and 51% similarity with the dog RAB8, mouse MEL, and mouse YPT1 proteins, respectively and contains the 4 GTP/GDP-binding sites that are present in all the RAS proteins. The putative effector-binding site of this protein is similar to that of the RAB/YPT proteins. However, this protein contains a C-terminal CAAX motif that is characteristic of many RAS superfamily members but which is not found in YPT1 and the majority of RAB proteins. Although this gene was isolated as a transforming gene from a melanoma cell line, no linkage between MEL and malignant melanoma has been demonstrable. This oncogene is located 800 kb distal to MY09B on chromosome 19p13.1.
Specifications
RAB8A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P55258, P61006 | |
RAB8A | |
human RAB8A recombinant protein. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Western Blot | |
261CT1.3.1 | |
Unconjugated | |
RAB8A | |
AA409338; cell line NK14 derived transforming oncogene; MEL; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); oncogene c-mel; Rab8; Rab8a; RAB8A, member RAS oncogene family; ras-associated protein RAB8; ras-related protein Rab-8A | |
Mouse | |
Protein G | |
RUO | |
17274, 4218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction