Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB8A Recombinant Rabbit Monoclonal Antibody (JE64-79)
GREENER_CHOICE

Catalog No. PIMA549140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549140 Supplier Invitrogen™ Supplier No. MA549140
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Sequence Similarities: 100% Mouse/Rat Tissue Specificity: Low tissue specificity Positive Control: Hela cell lysate, HepG2 cell lysate, NIH/3T3 cell lysate, RAW264.7 cell lysate, C2C12 cell lysate, L6 cell lysate Subcellular location: Cell membrane, golgi apparatus, recycling endosome membrane, cytoplasm, centriole, cilium basal body, cilium axoneme, cilium, phagosome, phagosome membrane, midbody.

The protein encoded by this gene is a member of the RAS superfamily which are small GTP/GDP-binding proteins with an average size of 200 amino acids. The RAS-related proteins of the RAB/YPT family may play a role in the transport of proteins from the endoplasmic reticulum to the Golgi and the plasma membrane. This protein shares 97%, 96%, and 51% similarity with the dog RAB8, mouse MEL, and mouse YPT1 proteins, respectively and contains the 4 GTP/GDP-binding sites that are present in all the RAS proteins. The putative effector-binding site of this protein is similar to that of the RAB/YPT proteins. However, this protein contains a C-terminal CAAX motif that is characteristic of many RAS superfamily members but which is not found in YPT1 and the majority of RAB proteins. Although this gene was isolated as a transforming gene from a melanoma cell line, no linkage between MEL and malignant melanoma has been demonstrable. This oncogene is located 800 kb distal to MY09B on chromosome 19p13.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB8A
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-79
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RAB8A
Gene Accession No. P35280, P55258, P61006
Gene Alias AA409338; cell line NK14 derived transforming oncogene; MEL; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); oncogene c-mel; Rab8; Rab8a; RAB8A, member RAS oncogene family; ras-associated protein RAB8; ras-related protein Rab-8A
Gene Symbols RAB8A
Host Species Rabbit
Immunogen Synthetic peptide within human rab8a aa 151-200/207.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117103, 17274, 4218
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.