Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567354 Supplier Invitrogen™ Supplier No. PA567354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSANVEEAFFT LARDIMTKLN RKMNDSNSAG AGGPVKITEN RSKKTSFFRC SLL Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab superfamilies exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. Rab 8B (Ras-related protein Rab-8B) is a 207 amino acid lipid anchor that localizes to the cytoplasmic side of the cell membrane and belongs to the Rab family and small GTPase superfamily. Rab 8B may be involved in neurotransmitter release and vesicular trafficking, and is encoded by a gene located on human chromosome Rab 8B and mouse chromosome 9 C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB8B
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB8B
Gene Accession No. Q92930
Gene Alias 5930437D16; D330025I23Rik; FLJ38125; GTPase Rab8b; LOW QUALITY PROTEIN: ras-related protein Rab-8B; RAB44, member RAS oncogene family; RAB8B; RAB-8b protein; RAB8B, member RAS oncogene family; rab8bb; ras-related protein Rab-8B; ras-related protein Rab-8B; LOW QUALITY PROTEIN: ras-related protein Rab-8B; Ras-related protein Rab-8B-like protein; Unknown (protein for MGC:127591); zgc:165529
Gene Symbols RAB8B
Host Species Rabbit
Immunogen Recombinant Human RAB8B. Recombinant protein control fragment (Product #RP-105589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.