Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB8B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727459 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7459 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7459 Supplier Proteintech Group Inc Supplier No. 117921AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab superfamilies exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. Rab 8B (Ras-related protein Rab-8B) is a 207 amino acid lipid anchor that localizes to the cytoplasmic side of the cell membrane and belongs to the Rab family and small GTPase superfamily. Rab 8B may be involved in neurotransmitter release and vesicular trafficking, and is encoded by a gene located on human chromosome Rab 8B and mouse chromosome 9 C.

Specifications

Antigen RAB8B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB8B
Gene Accession No. P61028, Q92930
Gene Alias RAB8, RAB8B, Ras related protein Rab 8B
Gene Symbols RAB8B
Host Species Rabbit
Immunogen RAB8B Fusion Protein Ag2421
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235442, 51762
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.