Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104086 Supplier Invitrogen™ Supplier No. PA5104086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB9B.

Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB9B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB9B
Gene Accession No. Q8BHH2, Q9NP90
Gene Alias 9330195C02Rik; Rab9b; RAB9B, member RAS oncogene family; RAB9L; Rab-9L; Rab-9-like protein; Ras-related protein Rab-9B; RGD1562951; si:dkey-169h1.8; TRAB9
Gene Symbols RAB9B
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB9B(Accession Q9NP90), corresponding to amino acid residues E104-S154.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319642, 367915, 51209
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.