Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB9A Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299423APC
Description
RAB9A Polyclonal antibody specifically detects RAB9A in Mouse samples. It is validated for Western BlotSpecifications
RAB9A | |
Polyclonal | |
Western Blot | |
RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB9A | |
0.1 mL | |
Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Signal Transduction | |
9367 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction