Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563992
Description
Immunogen sequence: EAVRQVLAVE EQLEHCMLGH TIDLNSGSKA GSSCC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
Specifications
RAB9B | |
Polyclonal | |
Unconjugated | |
RAB9B | |
9330195C02Rik; Rab9b; RAB9B, member RAS oncogene family; RAB9L; Rab-9L; Rab-9-like protein; Ras-related protein Rab-9B; RGD1562951; si:dkey-169h1.8; TRAB9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51209 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NP90 | |
RAB9B | |
Recombinant protein corresponding to Human Rab9. Recombinant protein control fragment (Product #RP-101985). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction