Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570252
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 79%; Zebrafish: 79%.
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
Specifications
RAB9B | |
Polyclonal | |
Unconjugated | |
RAB9B | |
9330195C02Rik; Rab9b; RAB9B, member RAS oncogene family; RAB9L; Rab-9L; Rab-9-like protein; Ras-related protein Rab-9B; RGD1562951; si:dkey-169h1.8; TRAB9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
319642 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BHH2 | |
RAB9B | |
synthetic peptide directed towards the middle region of human Rab9 (aa 58-107). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction