Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABAC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556078
Description
Immunogen sequence: MAAQKDQQKD AEAEGLSGTT LLPKLIPSGA GREWLERRRA TIRPWSTFVD QQRFSRP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Specifications
RABAC1 | |
Polyclonal | |
Unconjugated | |
RABAC1 | |
2310040I06Rik; Gbpap1; GTP binding protein associated protein 1; Pra; Pra1; PRA1 domain family 1; PRA1 family protein 1; Praf1; prenylated Rab acceptor 1; prenylated Rab acceptor protein 1; Prenylin; Rab acceptor 1; Rab acceptor 1 (prenylated); RABAC1; YIP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UI14 | |
RABAC1 | |
Recombinant protein corresponding to Human RABAC1. Recombinant protein control fragment (Product #RP-95702). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction