Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120688 Supplier Invitrogen™ Supplier No. PA5120688
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, U-251MG, SGC-7901, Mouse lung, Rat testis, Rat ovary. Immunogen sequence: MAAQKDQQKD AEAEGLSGTT LLPKLIPSGA GREWLERRRA TIRPWSTFVD QQRFSRPRNL GELCQRLVRN VEYYQSNYVF VFLGLILYCV VTSPMLLVAL AVFFGACYIL YLRTLESKLV LFGREVSPAH QYALAGGISF PFFWLAGAGS AVFWVLGATL VVIGSHAAFH QIEAVDGEEL QMEPV.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RABAC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RABAC1
Gene Accession No. O35394, Q9UI14, Q9Z0S9
Gene Alias 2310040I06Rik; Gbpap1; GTP binding protein associated protein 1; Pra; Pra1; PRA1 domain family 1; PRA1 family protein 1; Praf1; prenylated Rab acceptor 1; prenylated Rab acceptor protein 1; Prenylin; Rab acceptor 1; Rab acceptor 1 (prenylated); RABAC1; YIP3
Gene Symbols RABAC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human RABAC1 (NP_006414.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10567, 14470, 83583
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.