Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1700011F03Rik, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-384 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-384 Supplier Abnova Corporation Supplier No. PAB18825
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant 1700011F03Rik.

Specifications

Antigen 1700011F03Rik
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant 1700011F03Rik.
Dilution ELISA (1:5000-1:20000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene 1700011F03Rik
Gene Alias MGC58753
Gene Symbols 1700011F03Rik
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length mouse 1700011F03Rik.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74221
Target Species Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.