Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4932417H02Rik (Rptor), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116660 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-660 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-660 Supplier Abnova Corporation Supplier No. PAB13417
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Rptor.

Specifications

Antigen 4932417H02Rik (Rptor)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Rptor.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene 4932417H02Rik
Gene Alias Raptor/mKIAA1303
Gene Symbols 4932417H02Rik
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 16 amino acids of mouse Rptor.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74370
Test Specificity Raptor has multiple isoforms that may also be recognized by antibody.
Target Species Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.