Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AANAT, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89117412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-412 Supplier Abnova Corporation Supplier No. PAB15046

Rabbit polyclonal antibody raised against AANAT.

Arylalkylamine N-acetyltransferase belongs to the superfamily of acetyltransferases. It is the penultimate enzyme in melatonin synthesis and controls the night/day rhythm in melatonin production in the vertebrate pineal gland. Melatonin is essential for seasonal reproduction, modulates the function of the circadian clock in the suprachiasmatic nucleus, and influences activity and sleep. This enzyme is rapidly inactivated when animals are exposed to light at night. This protein is 80% identical to sheep and rat AA-NAT. Arylalkylamine N-acetyltransferase may contribute a multifactorial genetic diseases such as altered behavior in sleep/wake cycle. [provided by RefSeq

Specifications

Antigen AANAT
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against AANAT.
Dilution Western Blot (1:500-1:1000) Immunocytochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene AANAT
Gene Accession No. U40347
Gene Alias AA-NAT/SNAT
Gene Symbols AANAT
Host Species Rabbit
Immunogen Human AANAT.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15
Test Specificity Recognizes human and rhesus AANAT. It does not recognize rat, ovine, chicken or zebrafish AANAT. This is specific to primate AANAT (3,4). By Western blot, this antibody recognizes a band at 23 KDa representing hAANAT.
Target Species Human, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.