Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AARSD1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120088 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-088 Supplier Abnova Corporation Supplier No. PAB18528
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant AARSD1.

Specifications

Antigen AARSD1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant AARSD1.
Dilution Western Blot (1:1000-1:2000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene AARSD1
Gene Accession No. BC004172
Gene Alias MGC2744
Gene Symbols AARSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 278-522 of human AARSD1.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80755
Test Specificity This antibody is specific to AARSD1.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.