Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC4 Rabbit anti-Dog, Human, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116743 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-743 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-743 Supplier Abnova Corporation Supplier No. PAB13563

Rabbit polyclonal antibody raised against synthetic peptide of ABCC4.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms. [provided by RefSeq

Sequence: SGRLKEYDEPYVLLQNKESL

Specifications

Antigen ABCC4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ABCC4.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In borate buffer
Gene ABCC4
Gene Alias EST170205/MOAT-B/MOATB/MRP4
Gene Symbols ABCC4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1249-1268 of human ABCC4.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10257
Test Specificity Antibody binds cytochrome C.
Target Species Canine, Human, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.