Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCF3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-095 Supplier Abnova Corporation Supplier No. PAB18536
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant ABCF3.

Specifications

Antigen ABCF3
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant ABCF3.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene ABCF3
Gene Accession No. BC051754
Gene Alias EST201864/FLJ11198
Gene Symbols ABCF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 442-681 of human ABCF3.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55324
Test Specificity This antibody is specific to ABCF3.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.