Learn More
Specifications
Specifications
| Antigen | ABCG2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | In PBS (0.09% sodium azide) |
| Gene Accession No. | NP_004818;Q9UNQ0 |
| Gene Symbols | ABCG2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide (conjugated with KLH) corresponding to internal region of human ABCG2. |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.