Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-ABCG4, Polyclonal,

Catalog No. ABS1000MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1000MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1000MI Supplier MilliporeSigma™ Supplier No. ABS1000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects ABCG4 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry

ATP-binding cassette (ABC) sub-family G member 4 is a multi-membrane protein encoded by the ABCG4 (also known as WHITE2) gene in human. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White) with ABCG4 belonging to the White subfamily. ABCG4 is expressed primarily in the central nervous system in human and rat by Northern blot analysis (PMID 12183068). Transient expression in enterocytes and hematopoietic cells during murine embryonic development is also reported (PMID 19633360).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG4
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.
Gene Accession No. Q9H172
Gene Symbols ABCG4; WHITE2
Host Species Rabbit
Immunogen Linear peptide corresponding to the cytoplasmic domain of human ABCG4.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001135977
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.