Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121835 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-835 Supplier Abnova Corporation Supplier No. PAB20333
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ABHD2.

This gene encodes a protein containing an alpha/beta hydrolase fold, which is a catalytic domain found in a very wide range of enzymes. The function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding the same protein. [provided by RefSeq

Sequence: TALYGKMGRVRSPHPYGHRKFITMSDGATSTFDLFEPLAEHCVGDDITMVICPGIANHSEKQYIRTFVDYAQKNGYRCAVLNHLGALPNIELTSPRMFTYGCTWEFGAMVNYIKKTYPLTQLVVVGFSLGGNIVCKYLGETQANQEKVL

Specifications

Antigen ABHD2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ABHD2.
Dilution Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ABHD2
Gene Accession No. P08910
Gene Alias HS1-2/LABH2/MGC111112/MGC26249/PHPS1-2
Gene Symbols ABHD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ABHD2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11057
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.