Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAA1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-385 Supplier Abnova Corporation Supplier No. PAB18826
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant ACAA1.

This gene encodes an enzyme operative in the beta-oxidation system of the peroxisomes. Deficiency of this enzyme leads to pseudo-Zellweger syndrome. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen ACAA1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant ACAA1.
Dilution ELISA (1:10000-1:80000) Western Blot (1:200-1:1000) Immunohistochemistry (1:50-200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene ACAA1
Gene Accession No. NM_001607
Gene Alias ACAA/PTHIO/THIO
Gene Symbols ACAA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 210-424 of human ACAA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.