Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAD9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120081 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-081 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-081 Supplier Abnova Corporation Supplier No. PAB18521
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant ACAD9.

Mitochondrial fatty acid beta-oxidation is one of the main energy-producing metabolic pathways in eukaryotes. Acyl-CoA dehydrogenases (ACADs; EC 1.3.99.13) are mitochondrial enzymes that catalyze the initial rate-limiting step in the beta-oxidation of fatty acyl-CoA. ACAD9 belongs to a group of ACADs that act on fatty acids containing 14 to 20 carbons (Zhang et al., 2002 [PubMed 12359260]).[supplied by OMIM

Specifications

Antigen ACAD9
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant ACAD9.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene ACAD9
Gene Accession No. BC013354
Gene Alias ACAD-9/FLJ23533/MGC14452/NPD002
Gene Symbols ACAD9
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1464-1642 of human ACAD9.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28976
Test Specificity This antibody is specific to ACAD9.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.