Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-acetyl-Histone H3, Alexa Fluor 647, Polyclonal

Catalog No. 06599AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
06-599-AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-599-AF647 Supplier MilliporeSigma™ Supplier No. 06599AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects acetyl-Histone H3 in Human, Mouse samples, and it is validated for Immunocytochemistry

Histone H3 is one of the five main histone proteins that are involved in the structure of chromatin in eukaryotic cells. The chromatin fiber is composed on repetitive units, known as nucleosomes, which are comprised of an octamer of core histones (two each of H2A, H2B, H3, and H4), around which DNA are wrapped. Histone H3 features a main globular domain and a long N-terminal tail. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 is specifically phosphorylated during both mitosis and meiosis in patterns that are specifically coordinated in space and time. These phosphorylations are initiated at different phases of the cell division. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H3
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation Purified rabbit antibody conjugate in buffer containing PBS, 15mg/mL BSA and 0.1% Sodium Azide.
Gene Accession No. Q16695
Gene Symbols HIST3H3; H3FT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human histone H3 N-terminal sequence with acetylated Lys9 and Lys14
Purification Method Protein A Purfied
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.