Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-acetyl Histone H3 (Lys 64), Polyclonal,

Catalog No. ABE1057MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1057MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1057MI Supplier MilliporeSigma™ Supplier No. ABE1057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects acetyl Histone H3 (Lys 64) in Human, Mouse samples, and it is validated for Dot Blot

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci. Histone H3 is reported to be acetylated at Lys64 (H3K64ac) by p300 co-activator. In addition, H3K64ac is found enriched at the transcriptional start sites of active genes, consistent with a transcriptional activation modification opposite of its repressive counterpart H3K64me3 (PMID 24668167). The target specificity of ABE1057 is confirmed with peptide-based epitope analysis by dot blot, using AbSurance Histone H3 Antibody Specificity Array, and AbSurance Histone H2A, H2B, H4 Antibody Specificity Array. Sequence surrounding the target acetylation site is conserved among human Histone H3 subtypes H3.1, H3.2, H3.3, and H3.3C/H3.5 (UniProt P68431, Q71DI3, P84243, and Q6NXT2, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl Histone H3 (Lys 64)
Applications Dot Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P68431
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human acetyl Histone H3 (Lys 64).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003520
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.