Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACIN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134387 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-387 Supplier Abnova Corporation Supplier No. PAB9754

Rabbit polyclonal antibody raised against synthetic peptide of ACIN1.

Specifications

Antigen ACIN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description apoptotic chromatin condensation inducer 1
Formulation In PBS (0.02% sodium azide)
Gene ACIN1
Gene Alias ACINUS, ACN, DKFZp667N107, KIAA0670, fSAP152
Gene Symbols ACIN1
Host Species Rabbit
Immunogen A mixture of synthetic peptides corresponding to amino acids 775-789, 48-62, 17-31, 236-251 of human ACIN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22985
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.