Learn More
ACO1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACO1.
Supplier: Abnova Corporation PAB18048
Description
Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes. [provided by RefSeq]
Specifications
ACO1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ACO1. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P21399 | |
ACO1 | |
A synthetic peptide corresponding to human ACO1. | |
RUO | |
48 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50∼1:100) IELISA (1:20000) The optimal working dilution should be determined by the end user. | |
ACO1 | |
ACONS/IREB1/IREBP/IREBP1/IRP1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody is specific to ACO1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only