Learn More
ACOT13, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACOT13.
Supplier: Abnova Corporation PAB16788
Description
This gene encodes a member of the thioesterase superfamily. In humans, the protein co-localizes with microtubules and is essential for sustained cell proliferation. The orthologous mouse protein forms a homotetramer and is associated with mitochondria. The mouse protein functions as a medium- and long-chain acyl-CoA thioesterase. Multiple transcript variants encoding different isoforms have been found for this gene
Specifications
ACOT13 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ACOT13. | |
In PBS (0.02% sodium azide) | |
HT012/MGC4961/PNAS-27 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
THEM2 | |
THEM2 | |
A synthetic peptide corresponding to C-terminus 17 amino acids of human ACOT13. | |
RUO | |
55856 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only