Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL1 Rabbit anti-Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115871 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-871 Supplier Abnova Corporation Supplier No. PAB12497
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ACSL1.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. [provided by RefSeq

Specifications

Antigen ACSL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ACSL1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene ACSL1
Gene Accession No. P33121
Gene Alias ACS1/FACL1/FACL2/LACS/LACS1/LACS2
Gene Symbols ACSL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human ACSL1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2180
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.