Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSL6, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126733 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-733 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-733 Supplier Abnova Corporation Supplier No. PAB2506
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ACSL6.

Long-chain acyl-CoA synthetases (EC 6.2.1.3), such as ACSL6, catalyze the formation of acyl-CoA from fatty acids, ATP, and CoA (Malhotra et al., 1999 [PubMed 10548543]).[supplied by OMIM

Specifications

Antigen ACSL6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ACSL6.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ACSL6
Gene Accession No. ACSL6_HUMAN
Gene Alias ACS2/FACL6/FLJ16173/KIAA0837/LACS2/LACS5
Gene Symbols ACSL6
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of human ACSL6.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23305
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.