Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR1B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121266 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-266 Supplier Abnova Corporation Supplier No. PAB19738
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ACTR1B.

This gene encodes a 42.3kDa subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150kDa. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex. [provided by RefSeq

Specifications

Antigen ACTR1B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ACTR1B.
Dilution ELISA (1:160000) Western Blot (1:1000-2000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ACTR1B
Gene Accession No. P42025
Gene Alias ARP1B/CTRN2/PC3
Gene Symbols ACTR1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ACTR1B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10120
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.