Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACVRL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-310 Supplier Abnova Corporation Supplier No. PAB19782
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ACVRL1.

This gene encodes a type I cell-surface receptor for the TGF-beta superfamily of ligands. It shares with other type I receptors a high degree of similarity in serine-threonine kinase subdomains, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. The encoded protein, sometimes termed ALK1, shares similar domain structures with other closely related ALK or activin receptor-like kinase proteins that form a subfamily of receptor serine/threonine kinases. Mutations in this gene are associated with hemorrhagic telangiectasia type 2, also known as Rendu-Osler-Weber syndrome 2. [provided by RefSeq

Specifications

Antigen ACVRL1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ACVRL1.
Dilution ELISA (1:1000-1:10000) Western Blot (1:200-5000) Immunohistochemistry (1:100-300) The optimal working dilution should be determined by the end user.
Formulation In serum (2% BSA, 0.02% sodium azide)
Gene ACVRL1
Gene Accession No. NM_000020
Gene Alias ACVRLK1/ALK-1/ALK1/HHT/HHT2/ORW2/SKR3/TSR-I
Gene Symbols ACVRL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ACVRL1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.