Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADA (Bovine), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129580 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-580 Supplier Abnova Corporation Supplier No. PAB28071
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native ADA (Bovine).

Specifications

Antigen ADA (Bovine)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native ADA (Bovine).
Dilution ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene ADA
Gene Symbols ADA
Host Species Rabbit
Immunogen Native purified bovine ADA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280712
Target Species Bovine
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.